Learn More
HER-2/c-erbB-2/neu Ab-8 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Epredia™ HER-2/c-erbB-2/neu Ab-8, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-325-B0
Code : LD
Additional Details : Weight : 1.00000kg
Description
c-erbB-2 is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein. The c-erbB-2 protein is overexpressed in a variety of carcinomas especially those of breast and ovary.Host Species: Mouse
Clone: e2-4001
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: Cytoplasmic domain
Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein
Molecular Weight: 185 kDa
Positive Control: SKBR-3 cells, or T47D cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry
- Western Blotting
- Immunoprecipitation (Native and Denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
HER-2/c-erbB-2/neu Ab-8 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Oncogenesis | |
Human, Mouse, Rat | |
IgG1 |
Immunoprecipitation | |
e2-4001 | |
Biotin | |
Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein | |
RUO | |
Primary | |
Concentrated |