Learn More
Epredia™ Lab Vision™ HER-2/c-erB-2/neu Rabbit Monoclonal Antibody
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ HER-2/c-erB-2/neu, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
Brand: Epredia™ RM-9103-S
Description
c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. It is closely related in structure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a proportion of breast and other adenocarconomas, as well as transitional cell carcinomas. In the case of breast cancer, expression determined by immunohistochemistry has been shown to be associated with poor prognosis.Host Species: Rabbit
Clone: SP3
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Extracellular domain
Immunogen: Recombinant protein encoding extracellular domain of human HER-2
Molecular Weight: 185 kDa
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| HER-2/c-erB-2/neu | |
| Monoclonal | |
| Unconjugated | |
| Recombinant protein encoding extracellular domain of human HER-2 | |
| IVD | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| SP3 | |
| Rabbit | |
| 1 mL | |
| Oncogenesis | |
| Human | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.