missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1 Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1, Mouse Monoclonal Antibody.

£97.72 - £346.53

Specifications

Antigen Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1
Clone T9; same as 3E7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12620967
View Documents
Epredia™
MS-314-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12630967
View Documents
Epredia™
MS-314-S1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
£260.00
£243.32 / 500µL
Save £16.68
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12690957
View Documents
Epredia™
MS-314-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12600967
View Documents
Epredia™
MS-314-RQ
12 mL Ready-to-Use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12610967
View Documents
Epredia™
MS-314-S
1 mL Supernatant
ONLINE EXCLUSIVE PRICE
£368.00
£346.53 / 1mL
Save £21.47
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hepatitis B virus causes type B hepatitis and it belongs to hepadnavirus family. It is spherical in shape with a diameter of 42nm. It contains a 27nm partially double stranded DNA core enclosed within a lipoprotein coat. The antigenic activity of the nucleocapsid core is designated as hepatitis B core antigen. The antigens in the outer surface are called as hepatitis B virus surface antigens. Core antigens are localized within the nuclei whereas the surface antigens are present in the cytoplasm of the infected cells. Antibodies to surface antigens appear in circulation at an early stage of infection whereas the antibodies to the core antigens are detected after several weeks.

Host Species: Mouse

Clone: T9; same as 3E7

Isotype: IgG1/λ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified HBVsAg, subtype ayw3

Positive Control: Hepatitis B virus infected liver

Cellular Localization: Cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
ASR
Purified HbsAg, subtype ayw3
T9; same as 3E7
Monoclonal
Mouse
Infectious Disease
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.