missing translation for 'onlineSavingsMsg'
Learn More

GLUT-1 Rabbit Polyclonal Antibody, Epredia™

Product Code. 12653417 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Unit Size:
100µL
1mL
500µL
7mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
12653417 100 μL Concentrated 100µL
12663417 500 μL Concentrated 500µL
12643417 1 mL Concentrated 1mL
12683417 7 mL Prediluted 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12653417 Supplier Epredia Supplier No. RB9052P0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GLUT-1, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from C-terminal of human GLUT-1

Molecular Weight: 55 kDa, 45 kDa depending on the extent of glycosylation

Positive Control: Rat brain lysate; Breast CA, squamous cell CA, nonsmall-cell lung CA and esophageal CA

Cellular Localization: Membrane and focal cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide derived from C-terminal of human GLUT-1
Quantity 100 μL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.