Learn More
GLUT-1 (SPM498) Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ GLUT-1, Mouse Monoclonal Antibody.
£147.00 - £535.50
Specifications
| Antigen | GLUT-1 |
|---|---|
| Clone | SPM498 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12658786
|
Epredia
MS-10637-P0 |
100 μL | Concentrated | ||||||
|
12668786
|
Epredia
MS-10637-P1 |
500 μL | Concentrated | ||||||
|
12648786
|
Epredia
MS-10637-P |
1 mL | Concentrated | ||||||
|
12688786
|
Epredia
MS-10637-R7 |
7 mL | Prediluted | ||||||
Description
Glucose is fundamental to the metabolism in mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.Host Species: Mouse
Clone: SPM498
Isotype: IgG2a/κ
Species Reactivity: Human and Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from C-terminal of human GLUT-1
Molecular Weight: 55 kDa
Positive Control: Esophageal CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| GLUT-1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human, Rat | |
| IgG2a κ | |
| Esophageal CA |
| SPM498 | |
| Monoclonal | |
| Mouse | |
| Cancer and Tumor Biology | |
| A synthetic peptide derived from C-terminal of human GLUT-1 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.