missing translation for 'onlineSavingsMsg'
Learn More

Flt-1/VEGFR1 Ab-1 Rabbit Polyclonal Antibody, Epredia™

Ensure accurate, reproducible results in western blotting experiments with Epredia™ Flt-1/VEGFR1 Ab-1, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-1527-PABX

 View more versions of this product

Product Code. 12663037

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Flt-1/vascular endothelial growth factor receptor 1 (VEGFR1), a cell membrane receptor kinase, is a high-affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis. It contains seven immunoglobulin-like sequences in the extracellular domain and exhibits sequence similarity to Flk-1 and Flt-4.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin

Molecular Weight: 180 kDa

Positive Control: Hey cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flt-1/VEGFR1 Ab-1
Polyclonal
Unconjugated
Rabbit
1 mL
Angiogenesis
Human, Mouse
IgG
Western Blot
1mg/mL
Purified w/o BSA and Azide
A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.