missing translation for 'onlineSavingsMsg'
Learn More

Epithelial Membrane Antigen/EMA Ab-3 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Membrane Antigen/EMA Ab-3, Mouse Monoclonal Antibody.

Brand:  Epredia MS-741-P0

Code : LD

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12662207

  • £105.00 / 0.10mL
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Epithelial membrane antigen (EMA), is a mucin-like glycoprotein. Antibody to EMA has been shown useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in the bone marrow or liver.

Host Species: Mouse

Clone: E29

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: RPAP

Immunogen: Delipidated extract of human milk fat globule membranes

Molecular Weight: 265-400 kDa

Positive Control: Breast or Colon CA

Cellular Localization: Cytoplasmic and Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Epithelial Membrane Antigen/EMA Ab-3
Monoclonal
200μg/mL
Mouse
100 μL
Oncogenesis
Human
IgG2a κ
Immunohistochemistry (Paraffin)
E29
Unconjugated
Delipidated extract of human milk fat globule membranes
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers