missing translation for 'onlineSavingsMsg'
Learn More

Epidermal Growth Factor Receptor/EGFR Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunoprecipitation experiments with Epredia™ Epidermal Growth Factor Receptor/EGFR Ab-2, Mouse Monoclonal Antibody.

£136.00 - £515.10

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-2
Clone 225
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12610757
View Documents
Epredia
MS-269-P0
100 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12620757
View Documents
Epredia
MS-269-P1
500 μL 200μg/mL
ONLINE EXCLUSIVE PRICE
£353.00
£332.24 / 500µL
Save £20.76
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12600757
Ver documentos
Epredia
MS-269-P
1 mL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
Ahorre hasta 
»
12630757
Ver documentos
Epredia
MS-269-P1ABX
100 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
Ahorre hasta 
»
12640757
Ver documentos
Epredia
MS-269-PABX
200 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
Ahorre hasta 
»
Descripción

Descripción

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 225

Isotype: IgG1

Species Reactivity: Human, Chimpanzee and Monkey. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: Purified EGFR from A431 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Immunoprecipitation (Native only)
  • Blocks EGF/TGFα-Induced Activation of EGFR
  • Arrests Tumor Growth in vivo
  • Inhibits Binding of Ab-1 (clone 528) to EGFR

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Epidermal Growth Factor Receptor/EGFR Ab-2
Immunoprecipitation
Unconjugated
Mouse
EGFR Family
Purified EGFR from A431 cells
Primary
225
Monoclonal
Concentrated
RUO
Human, Non-human Primate
IgG1
Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado