missing translation for 'onlineSavingsMsg'
Learn More

Dystrophin Rabbit Polyclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Dystrophin, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-9024-R7

 View more versions of this product

Product Code. 12623347

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immuohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide corresponding to C-terminal of human Dystrophin

Molecular Weight: 427 kDa

Positive Control: Skeletal muscle

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dystrophin
Polyclonal
Rabbit
7 mL
Muscle Biology
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
A synthetic peptide corresponding to C-terminal of human Dystrophin
IVD
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.