missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ DP-1 Transcription Factor Ab-6 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in western blotting experiments with Epredia™ DP-1 Transcription Factor Ab-6, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1056-PABX

 View more versions of this product

Product Code. 12638726

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

DP family contains at least two members, DP-1 and -2. Structurally, DP-1 and -2 are relatively conserved, yet their expression appears to be tissue specific. E2F-1, a functional target for the tumor suppressor protein Rb, heterodimers with DP-1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.

Host Species: Mouse

Clone: 1DP06 (TFD10)

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 83-204

Immunogen: Recombinant DP-1 protein

Molecular Weight: 45 kDa

Positive Control: MAD109 or Raji cells, Tonsil

Cellular Localization: Cytoplasm and nucleus

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DP-1 Transcription Factor Ab-6
Monoclonal
1mg/mL
Mouse
200 μL
Transcription Regulation
Human, Mouse, Rat
IgG2a κ
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1DP06; same as TFD10
Unconjugated
Recombinant DP-1 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.