missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 10 Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Brand:  Epredia MS-611-P1ABX

 View more versions of this product

Product Code. 12611767

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CK10 (Cytokeratin 10) is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of CK10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. Expression of CK10 is related to maturation of malignant keratinocytes, being preferentially detected in more-differentiated parts.

Host Species: Mouse

Clone: DE-K10

Isotype: IgG1

Species Reactivity: Human, Rat, Dog and Cat. Others not known.

Epitope: Not determined

Immunogen: Cytoskeletal preparation extracted from human ectocervical epithelium

Molecular Weight: 56.5 kDa

Positive Control: A431 cells, Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 10 Ab-2
Monoclonal
1mg/mL
Mouse
100 μL
Cytoskeletal Protein
Canine, Feline, Human, Rat
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
DE-K10
Unconjugated
Cytoskeletal preparation extracted from human ectocervical epithelium
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.