missing translation for 'onlineSavingsMsg'
Learn More

Cyclin D2 Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry experiments.

Brand:  Epredia MS-213-P1ABX

 View more versions of this product

Product Code. 12660567

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Cyclin D2 is a G1 cyclin required for G1-phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk4 and/or cdk6.

Host Species: Mouse

Clone: DCS-3.1

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human recombinant full-length cyclin D2 protein

Molecular Weight: 34 kDa

Positive Control: U-2-OS, C82, RD, Bristol-8 or LoVo36 cells. Note that T47D, ZR75, SKBR3, MCF-7, BT-20, MDA468, HBL100, BT5549, HeLa, Saos2 and HT29 cells are negative for cyclin D2.

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Biological Blockade
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclin D2 Ab-2
Monoclonal
1mg/mL
Mouse
100 μL
Cell Cycle
Human, Mouse, Rat
IgG2a κ
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
DCS-3.1
Unconjugated
Purified human recombinant full length cyclin D2 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.