Learn More
Epredia™ Lab Vision™ CEA (Carcinoembryonic Antigen)/CD66e Ab-3 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.
£132.25 - £398.68
Specifications
| Antigen | CEA (Carcinoembryonic Antigen)/CD66e Ab-3 |
|---|---|
| Clone | COL-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|
12691777
|
Epredia™
MS-613-B1 |
500 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12671777
|
Epredia™
MS-613-B |
1 mL | Biotin | Concentrated | 200μg/mL | ||||
|
12681787
|
Epredia™
MS-613-R7 |
7 mL | Unconjugated | Prediluted | |||||
Description
Host Species: Mouse
Clone: COL-1
Isotype: IgG2a/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Immunogen: Human colon carcinoma extract
Molecular Weight: 180 kDa
Positive Control: LS174T cells, Colon CA
Cellular Localization: Cytoplasmic and luminal membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
| CEA (Carcinoembryonic Antigen)/CD66e Ab-3 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Mouse | |
| Oncogenesis | |
| IgG2a κ |
| COL-1 | |
| Monoclonal | |
| RUO | |
| Human | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.