Learn More
Epredia™ Lab Vision™ Cdh1 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Epredia™ Cdh1 Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1116-PABX
Description
Research Areas: Cell Cycle; Cdh1 protein is a limiting, substrate-specific activator of anaphase-promoting complex (APC)-dependent proteolysis and it is required for the degradation of the APC substrates, such as Clb2 and Ase1. Cdh1 plays important role in mitotic regulation.Host Species: Mouse
Clone: DH01; same as DCS-266
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human Cdh1 protein
Molecular Weight: ∽55 kDa
Positive Control: Jurkat or LS174T cells
Recommended for:
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
| Cdh1 Ab-1 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 200 μL | |
| Cell Cycle | |
| Human | |
| IgG2a κ |
| Immunoprecipitation, Western Blot | |
| DH01; same as DCS-266 | |
| Unconjugated | |
| Recombinant human cdh1 protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.