missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD8 Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD8 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-457-R7

Code : LD

Additional Details : Weight : 0.01200kg

 View more versions of this product

Product Code. 12611497

  • £322.00 / 7mL
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD8 molecule consists of two chains, termed α and Β chain, which are expressed as a disulphide-α β heterdimer or as an alpha α homodimer on T cell subset (cytotoxic/suppressor T cells), thymocytes and NK cells. The majority of CD8+ Tcells expresses CD8 as αβheterodimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 α chain (but not Β chain).

Host Species: Mouse

Clone: C8/144B

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: C-terminal

Immunogen: A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of α-chain of human CD8 molecule

Molecular Weight: 32 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD8 Ab-1
Monoclonal
Unconjugated
A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of alpha chain of human CD8 molecule
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
C8/144B
Mouse
7 mL
T-Cell
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers