missing translation for 'onlineSavingsMsg'
Learn More

CD79a/mb-1 (B-Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Brand:  Epredia MS-357-R7

 View more versions of this product

Product Code. 12691167

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.

Host Species: Mouse

Clone: HM47/A9

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Horse, Cow, Pig, Rabbit, Guinea Pig, Rat, Mouse and Chicken (Plasma cells). Does not react with opossum.

Epitope: GTYQDVGSLNIADVQ

Immunogen: A synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein

Molecular Weight: 47 kDa

Positive Control: Raji cells, Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD79a/mb-1 (B-Cell Marker) Ab-1
Monoclonal
Prediluted
Mouse
7 mL
B-Cell
Bovine, Chicken, Equine, Guinea Pig, Human, Mouse, Non-human Primate, Pig, Rabbit, Rat
IgG2a κ
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
HM47/A9
Unconjugated
A synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.