missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD45RA Ab-3 Mouse Monoclonal Antibody, 1mL; 200μg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD45RA Ab-3, Mouse Monoclonal Antibody.

£370.26

Specifications

Antigen CD45RA Ab-3
Clone 4KB5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12610277
View Documents
Epredia™
MS-1845-P
1 mL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

CD45 or Leukocyte Common Antigen (LCA/T200) belongs to a family of membrane sialoglycoproteins with MW of 180kDa-220 kDa. Human CD45 contains three exons which encode peptide segments designated A, B and C, respectively. The differential splicing of the exons generates various isoforms. CD45RA.is a single chain glycoprotein. It is expressed on appoximately 40-50% of peripheral CD4+ T-cells, 50% of peripheral CD8+ T-cells, B-cells, and leukemic B-cell lines. T-cells expressing CD45RA and CD45RO define complementary, predominantly non-overlapping populations of resting peripheral T-cells. CD45RA are naive or virgin T-cells while, T-cells expressing CD45RO are memory T-cells.

Host Species: Mouse

Clone: 4KB5

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested

Epitope: Not determined

Immunogen: Hairy cell leukemia cells

Molecular Weight: 205-220 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD45RA Ab-3
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgG2a κ
4KB5
Monoclonal
Mouse
Oncogenesis
Hairy cell leukemia cells
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.