Learn More
CD45/T200/LCA Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, and flow cytometry experiments.
Brand: Epredia MS-355-RQ
Description
CD45 leucocyte common antigen (LCA) belongs to the family of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues. The intracellular portion of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals. Antibody to CD45 is useful in differential identification of lymphoid tumors from non-hematopoietic undifferentiated neoplasms.Host Species: Mouse
Clone: PD7/26/16 + 2B11
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat and dog. Others not known
Epitope: Not determined
Immunogen: PD7/26/16: human peripheral blood lymphocytes maintained in T-cell growth factor. 2B11: isolated neoplastic cells from T-cell lymphoma
Molecular Weight: 220, 205, 190, 180 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
- Flow Cytometry
Specifications
| CD45/T200/LCA Ab-2 | |
| Monoclonal | |
| Prediluted | |
| Mouse | |
| 12 mL | |
| Oncogenesis | |
| Ready-to-Use |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PD7/26/16 + 2B11 | |
| Unconjugated | |
| PD7/26/16: human peripheral blood lymphocytes maintained in T cell growth factor. 2B11: isolated neoplastic cells from T cell lymphoma. | |
| IVD | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.