Learn More
Epredia™ Lab Vision™ CD138 Rabbit Polyclonal Antibody with BSA and azide
Ensure accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD138, Rabbit Polyclonal Antibody.
Brand: Epredia™ RB-9422-P0
Description
CD138/Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is a selective marker for B cell lymphoblastic leukemia and lymphoplasmocytoid leukemia. It is lost from the apoptotic myeloma cells; hence is a useful marker for viable myeloma cells.Host Species: Rabbit
Species Reactivity: Human. Others not tested
Epitope: aa 18-218
Immunogen: Recombinant protein encoding aa 18-218 human CD138
Molecular Weight: 30.5kDa (core protein); 85-92kDa (depending upon glycosaminoglycans)
Positive Control: Tonsil
Cellular Localization: Plasma cell and Membrane
Recommended for:
- Immunohistochemistry
Note:
USA: RUO; Int'l: RUO
Specifications
| CD138 | |
| Polyclonal | |
| Unconjugated | |
| Recombinant protein encoding aa 18-218 human CD138 | |
| RUO | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| 200µg/mL | |
| Rabbit | |
| 100 μL | |
| Cell Adhesion | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.