Learn More
Epredia™ Lab Vision™ Caspase 6 (Mch 2) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and Azide
Ensure accurate, reproducible results in western blotting and immunoprecipitation procedures with Epredia™ Caspase 6 (Mch 2) Ab-3, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1126-P1
Code : LD
Additional Details : Weight : 1.00000kg
Description
Caspases play a major role in the transduction of the apoptotic signal and execution of apoptosis in mammalian cells. Caspase 3 (CPP32) and Caspase 6 (Mch2) are the major active caspases in apoptotic cells, and are activated in response to distinct apoptosis-inducing stimuli in all cell lines analyzed. Both CPP32 and Mch2 are present in apoptotic cells as multiple active species. Caspase-6 cleaves nuclear mitotic apparatus protein (NuMA) and mediates the shrinkage and fragmentation of nuclei.Host Species: Mouse
Clone: 6CSP03; same as 14.1.190
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Caspase 6 prodomain
Immunogen: Recombinant human caspase 6 prodomain
Molecular Weight: 34 kDa
Positive Control: Jurkat cells
Cellular Localization: Cytoplasmic
Recommended for:
- Western Blotting
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
Caspase 6 (Mch 2) Ab-3 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Apoptosis | |
Human | |
IgG2a κ |
Immunoprecipitation, Western Blot | |
6CSP03; same as 14.1.190 | |
Unconjugated | |
Recombinant human caspase 6 prodomain | |
RUO | |
Primary | |
Concentrated |