missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Caldesmon, HMW, Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Caldesmon, HMW Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1169-PABX

 View more versions of this product

Product Code. 12689186

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their eletrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.

Host Species: Mouse

Clone: h-CALD

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 150kDa (h-caldesmon)

Positive Control: Uterus

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Caldesmon, HMW Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Muscle Biology
Human
IgG2a κ
Immunohistochemistry (Paraffin)
h-CALD
Unconjugated
Crude human uterus extract
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.