Learn More
Epredia™ Lab Vision™ Caldesmon, HMW, Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Caldesmon, HMW Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1169-P1
Description
Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their eletrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.Host Species: Mouse
Clone: h-CALD
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Crude human uterus extract
Molecular Weight: 150kDa (h-caldesmon)
Positive Control: Uterus
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Caldesmon, HMW Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Muscle Biology | |
| Human | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| h-CALD | |
| Unconjugated | |
| Crude human uterus extract | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.