missing translation for 'onlineSavingsMsg'
Learn More

c-myc Ab-2 Mouse Monoclonal Antibody, Epredia™

Epredia™ c-myc Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-139-P

 View more versions of this product

Product Code. 12699756

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors.

Host Species: Mouse

Clone: 9E10.3

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse, rat and chicken. Others not tested.

Epitope: aa 410-419

Immunogen: A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc

Molecular Weight: 64-67 kDa

Positive Control: Raji cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry
  • Electron Microscopy
  • Affinity Purification, Immunoprecipitation and Western Blotting of fusion proteins labeled with EQKLISEEDL tag

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

c-myc Ab-2
Monoclonal
200μg/mL
Mouse
1 mL
Transcription Regulation
Human
IgG2a κ
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
9E10.3
Unconjugated
A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.