missing translation for 'onlineSavingsMsg'
Learn More

c-myc Ab-2 Mouse Monoclonal Antibody, Epredia™

Product Code. 12689756 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
Unit Size:
100µL
1mL
200µL
500µL
7mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conjugate Form Concentration unitSize
12689756 500 μL Biotin Concentrated 200μg/mL 500µL
12679756 100 μL Biotin Concentrated 200μg/mL 100µL
12609766 100 μL Unconjugated Concentrated 200μg/mL 100µL
12619766 100 μL Unconjugated Concentrated 1mg/mL 100µL
12639766 200 μL Unconjugated Concentrated 1mg/mL 200µL
12629766 500 μL Unconjugated Concentrated 200μg/mL 500µL
12699756 1 mL Unconjugated Concentrated 200μg/mL 1mL
12679766 7 mL Unconjugated Prediluted Prediluted 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 12689756 Supplier Epredia Supplier No. MS139B1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Epredia™ c-myc Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors.

Host Species: Mouse

Clone: 9E10.3

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse, rat and chicken. Others not tested.

Epitope: aa 410-419

Immunogen: A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc

Molecular Weight: 64-67 kDa

Positive Control: Raji cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry
  • Electron Microscopy
  • Affinity Purification, Immunoprecipitation and Western Blotting of fusion proteins labeled with EQKLISEEDL tag

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen c-myc Ab-2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9E10.3
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc
Quantity 500 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.