missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Bcl-2α Ab-3 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments.

Brand:  Epredia™ MS-597-B0

 View more versions of this product

Product Code. 12691707

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.

Host Species: Mouse

Clone: 8C8

Isotype: IgG1

Species Reactivity: Human and Monkey. Does not react with mouse and rat. Others not tested.

Epitope: aa 41-54

Immunogen: A synthetic peptide, aa 41-54 (Cys-GAAPAPGIFSSQPG) of human Bcl-2 protein

Molecular Weight: 25-26kDa

Positive Control: Raji or HeLa Cells, Tonsil or follicular lymphomas

Cellular Localization: Cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bcl-2alpha Ab-3
Monoclonal
200μg/mL
Mouse
100 μL
Apoptosis
Human, Non-human Primate
IgG1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
8C8
Biotin
A Synthetic peptide, aa 41-54 (Cys-GAAPAPGIFSSQPG) of human bcl-2 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.