missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Autostainer 480S-2D for 48 slides with 2D barcode reader and PT module 240V

Product Code. 15869452 Shop All Epredia Products
Change view
Click to view available options
Slide Capacity:
1 to 48 Slides/Run
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Slide Capacity Unit Size
15869452 1 to 48 Slides/Run Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Products on this page may include voltages not considered standard in North America. Please review carefully.

Product Code. 15869452 Supplier Epredia™ Supplier No. A80500027
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Products on this page may include voltages not considered standard in North America. Please review carefully.

Stain at least 108 slides per day with this fast, space-saving unit that provides flexible options to meet the workflow needs of any laboratory.

Benefits:
  • One to 48 slide capacity, each individually programmable
  • User-specified protocols and choice of reagents
  • Multiple staining runs per day
  • Truly hands-free, walk-away operation with advanced timer functions for overnight use
  • Easy, Windows™ 7 based programming
  • Modular options with up to four units controlled from one PC
  • Complemented by the Epredia™ PT Module designed for fully automated, simultaneous dewaxing and epitope recovery (The PT Module is available separately or in a system package).
  • Fast flow software logic enables flexible programming, operator ease of use and control of IHC costs

System Features:

  • Incorporates 2D Data Matrix Bar Coding for Slides and Reagents
  • Can be configured for research or diagnostic applications at no additional cost
  • Scans rapidly using high precision label detector technology 
  • Has integrated digital bar code scanner
  • Fully configured 2D data matrix software and printer interface
  • Bar code control of all protocol, reagent and slide operations
  • Preprintable data matrix labels with user-defined autoprograms
  • Adaptable for proprietary 2D-labeled reagents or user-defined reagents/labels
  • Optional individual inactivation of slide and/or reagent bar coding

Includes:

  • 48-slide Autostainer with 2D matrix bar code reader
  • Computer
  • Monitor
  • Label printer(s)
  • Document printer
  • Three metal reinforced, ABS plastic slide racks
  • Reagent rack Reagent vials and caps
  • Autostainer probe cleaning reagent kit
  • Buffer rinse bottle
  • Wash Buffer, DI water and waste containers
  • Universal Power Supply
TRUSTED_SUSTAINABILITY

Specifications

Description Autostainer 480S + PT Module 240V
Slide Capacity 1 to 48 Slides/Run
Carryover <10-6
Reagent Capacity Up to 40 reagents/run, 12mL/reagent
Temperature Range (Metric) Ambient
Weight (Metric) ∼54 kg
Height (Metric) 58 cm
Length (Metric) 67 cm
Width (Metric) 76 cm
Voltage 240 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.