missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Antibody Diluent OP Quanto
Shop All Epredia Products
Click to view available options
Quantity:
125 mL
Unit Size:
Each
Description
- Contains a protein stabilizing agent and preservative
- Suitable for use as a negative control reagent during immunohistochemical staining
- Concentrated antibodies can be diluted to the appropriate titer specified on the individual antibody datasheet or determined by the user, to achieve optimal staining
Specifications
Specifications
| Capacity | 125 mL |
| Content And Storage | 2° to 8°C |
| Product Type | Antibody Diluent |
| Quantity | 125 mL |
For in vitro diagnostic use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction