missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Contains a protein stabilizing agent and preservative
- Suitable for use as a negative control reagent during immunohistochemical staining
- Concentrated antibodies can be diluted to the appropriate titer specified on the individual antibody datasheet or determined by the user, to achieve optimal staining
Specifications
Specifications
| Capacity | 125 mL |
| Content And Storage | 2° to 8°C |
| Product Type | Antibody Diluent |
| Quantity | 125 mL |
For in vitro diagnostic use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?