Learn More
Berthold Technologies Lab Jack With Table 200 x 200 mm
Product Code. 31777305 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Imaging System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31777305 Imaging System Each
1 options
Product Code. 31777305 Supplier Berthold Technologies Supplier No. 64469
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Used in the NightSHADE imaging system to bring the samples closer to the camera.
  • The camera position in the Berthold NightSHADE in vivo imaging system is fixed
  • The lab jack raises the sample closer to the camera
  • Supports imaging of small samples
  • Allows imaging of specific areas of larger samples
Specifications
Type Support Stand
For Use With (Equipment) Imaging System
Depth (Metric) 200 mm
Length (Metric) 200 mm