Learn More
Cole-Parmer™ Lab-Crest™ Neoprene Needle Valve O-Ring
Shop All Cole-Parmer Products
Click to view available options
:
For use with: Lab-Crest Pressure Reaction Vessels
Description
- Parts and accessories for your Lab-Crest system.
- Needle valve provides flow control of gases at inlet pressures up to 2000psi.
Specifications
Specifications
| Description | Needle Valve O-Ring |
| Material | Neoprene |
| For Use With | Lab-Crest Pressure Reaction Vessels |