Learn More
Invitrogen™ La Crosse Virus Monoclonal Antibody (10G5.4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110804
Description
MA1-10804 detects La Crosse Virus and recognizes G1 and G2. MA1-10804 has been successfully used in ELISA, Western blot, Immunofluorescence, immunoprecipitation, and neutralization applications. The MA1-10804 immunogen is La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation.
Specifications
| La Crosse Virus | |
| Monoclonal | |
| 1.3 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Immunoprecipitation, Neutralization, Western Blot, Immunocytochemistry | |
| 10G5.4 | |
| Unconjugated | |
| LCV | |
| La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.