missing translation for 'onlineSavingsMsg'
Learn More

L755507, MedChemExpress

Product Code. 16470198
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470198 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16470198 Supplier Medchem Express Supplier No. HY19334
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress L755507 is a potent, selective agonist of β3-AR with an IC50 of 35 nM. L755507 enhances the homology-directed repair (HDR)-mediated genome editing in CRISPR/Cas9 nickase system.

L755507 is a potent, selective agonist of β3-AR with an IC50 of 35 nM. L755507 enhances the homology-directed repair (HDR)-mediated genome editing in CRISPR/Cas9 nickase system.

Specifications

Chemical Name or Material L755507
CAS 159182-43-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H40N4O6S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (171.02 mM)
SMILES O=S(C1=CC=C(NC(NCCCCCC)=O)C=C1)(NC2=CC=C(CCNC[C@H](O)COC3=CC=C(O)C=C3)C=C2)=O
Molecular Weight (g/mol) 584.73
Formula Weight 584.73
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.