missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ L3MBTL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584800
This item is not returnable.
View return policy
Description
Immunogen sequence: YWCDVNSPYV QPVGWCQENG RTLIAPQGYP NPENFSWTEY LEATQTNAVP AKVFKMRLPH GFL Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 59%.
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
Specifications
| L3MBTL4 | |
| Polyclonal | |
| Unconjugated | |
| L3MBTL4 | |
| A730037L19Rik; D930040M24Rik; H-l(3)mbt-like protein 4; HsT1031; l(3)mbt-like 4 (Drosophila); l(3)mbt-like protein 4; L3MBTL4; Lethal(3)malignant brain tumor-like protein 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91133 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NA19 | |
| L3MBTL4 | |
| Recombinant protein corresponding to Human L3MBTL4. Recombinant protein control fragment (Product #RP-107669). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction