missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14070-1-AP-20UL
This item is not returnable.
View return policy
Description
Ran/TC4-binding protein, RanBP1, interacts specifically with GTP-charged RAN. RANBP1 encodes a 23-kD protein that binds to RAN complexed with GTP but not GDP. RANBP1 does not activate GTPase activity of RAN but does markedly increase GTP hydrolysis by the RanGTPase-activating protein (RanGAP1). The RANBP1 cDNA encodes a 201-amino acid protein that is 92% similar to its mouse homolog. In both mammalian cells and in yeast, RANBP1 acts as a negative regulator of RCC1 by inhibiting RCC1-stimulated guanine nucleotide release from RAN.Specifications
| L3MBTL3 | |
| Polyclonal | |
| Unconjugated | |
| L3MBTL3 | |
| H l(3)mbt like protein 3, KIAA1798, l(3)mbt like 3 (Drosophila), L(3)mbt like protein 3, L3MBTL3, MBT 1, MBT1, RP11 73O6.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 237339, 84456 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BLB7, Q96JM7 | |
| L3MBTL3 | |
| L3MBTL3 Fusion Protein Ag5130 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction