Learn More
L3MBTL2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24435-1-AP-20UL
This item is not returnable.
View return policy
Description
The L3MBTL2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human L3MBTL2. This antibody recognizes human antigen. The L3MBTL2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| L3MBTL2 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| H l(3)mbt l, H l(3)mbt like protein 2, l(3)mbt like 2 (Drosophila), L(3)mbt like protein 2, L3MBT, L3MBTL2 | |
| 20 μL | |
| 83746 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |