missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | L3MBTL2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18323367
|
Novus Biologicals
NBP3-05067-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18335806
|
Novus Biologicals
NBP3-05067-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L3MBTL2 Polyclonal specifically detects L3MBTL2 in Human samples. It is validated for Western Blot.Specifications
| L3MBTL2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| dJ756G23.3, DKFZP761I141, H-l(3)mbt-l, H-l(3)mbt-like protein 2, l(3)mbt-like 2 (Drosophila), L(3)mbt-like protein 2, L3MBT, lethal(3)malignant brain tumor-like protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 486-705 of human L3MBTL2 (NP_113676.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS with 50% glycerol, pH7.3. | |
| 83746 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title