missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
L3MBTL2 Polyclonal specifically detects L3MBTL2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | L3MBTL2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | dJ756G23.3, DKFZP761I141, H-l(3)mbt-l, H-l(3)mbt-like protein 2, l(3)mbt-like 2 (Drosophila), L(3)mbt-like protein 2, L3MBT, lethal(3)malignant brain tumor-like protein 2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human L3MBTL2 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?