missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ L3MBTL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115488
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total L3MBTL1.
L3MBTL1 is the homolog of a protein identified in Drosophila as a suppressor of malignant transformation of neuroblasts and ganglion-mother cells in the optic centers of the brain. L3MBTL1 is localized to condensed chromosomes in mitotic cells and overexpression of this protein in a glioma cell line results in improper nuclear segregation and cytokinesis producing multinucleated cells. L3MBTL1 represents a polycomb group gene the encoded protein functions to regulate gene activity, likely via chromatin modification. Polycomb group protein specifically recognizes and binds mono- and dimethyllysine residues on target proteins, thereby acting as a reader of a network of post-translational modifications. Alternatively spliced transcript variants encoding different isoforms of the L3MBTL1 gene have been identified.
Specifications
| L3MBTL1 | |
| Polyclonal | |
| Unconjugated | |
| L3MBTL1 | |
| C630004G01; dJ138B7.3; H-l(3)mbt; H-l(3)mbt protein; Kiaa0681; L(3)mbt protein homolog; l(3)mbt-like (Drosophila); l(3)mbt-like 1; l(3)mbt-like 1 (Drosophila); L3mbt; L3MBTL; L3MBTL histone methyl-lysine binding protein 1; L3MBTL1; L3MBTL1, histone methyl-lysine binding protein; lethal (3) malignant brain tumor l(3); lethal(3)malignant brain tumor-like protein 1; lethal(3)malignant brain tumor-like protein 1; serine/threonine-protein kinase Sgk2; LOC100070683; mKIAA0681; RGD1307316; RP1-138B7.1; ZC2HC3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 241764, 26013, 311613 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| A2A5N8, D3ZWK4, Q9Y468 | |
| L3MBTL1 | |
| A synthesized peptide derived from human L3MBTL1(Accession Q9Y468), corresponding to amino acid residues D452-S502. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction