missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ L3HYPDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546309
This item is not returnable.
View return policy
Description
Peptide sequence: GTILTDGKDA YTKEPTTNIC VFADEQVDRS PTGSGVTARI ALQYHKGLLE.
L3HYPDH catalyzes the dehydration of trans-3-hydroxy-L-proline to delta-1-pyrroline-2-carboxylate (Pyr2C). It may be required to degrade trans-3-hydroxy-L-proline from the diet and originating from the degradation of proteins such as collagen-IV that contain it.
Specifications
| L3HYPDH | |
| Polyclonal | |
| Unconjugated | |
| L3HYPDH | |
| C14orf149; L-3-hydroxyproline dehydratase (trans-); L3HYPDH; probable proline racemase; trans-3-hydroxyl-L-proline dehydratase; trans-3-hydroxy-L-proline dehydratase; trans-L-3-hydroxyproline dehydratase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 112849 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96EM0 | |
| L3HYPDH | |
| Synthetic peptide directed towards the C-terminal of human L3HYPDH (aa 242-291). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction