missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ L3HYPDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563342
This item is not returnable.
View return policy
Description
Immunogen sequence: MESALAVPRL PPHDPGTPVL SVVDMHTGGE PLRIVLAGCP EVSGPTLLAK RRYMRQHLDH VRRRLMFEPR GHRDMYGAVL VPSEL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
L3HYPDH catalyzes the dehydration of trans-3-hydroxy-L-proline to delta-1-pyrroline-2-carboxylate (Pyr2C). It may be required to degrade trans-3-hydroxy-L-proline from the diet and originating from the degradation of proteins such as collagen-IV that contain it.
Specifications
| L3HYPDH | |
| Polyclonal | |
| Unconjugated | |
| L3HYPDH | |
| C14orf149; L-3-hydroxyproline dehydratase (trans-); L3HYPDH; probable proline racemase; trans-3-hydroxyl-L-proline dehydratase; trans-3-hydroxy-L-proline dehydratase; trans-L-3-hydroxyproline dehydratase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112849 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96EM0 | |
| L3HYPDH | |
| Recombinant protein corresponding to Human L3HYPDH. Recombinant protein control fragment (Product #RP-101260). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction