missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L2HGDH Mouse anti-Human, Porcine, Clone: 3D11C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66269-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes L-2-hydroxyglutarate dehydrogenase, a FAD-dependent enzyme that oxidizes L-2-hydroxyglutarate to alpha-ketoglutarate in a variety of mammalian tissues. Mutations in this gene cause L-2-hydroxyglutaric aciduria, a rare autosomal recessive neurometabolic disorder resulting in moderate to severe mental retardation.Specifications
| L2HGDH | |
| Monoclonal | |
| 1.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H9P8 | |
| L2HGDH | |
| L2HGDH Fusion Protein Ag8382 | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3D11C2 | |
| Unconjugated | |
| L2HGDH | |
| C14orf160, DURANIN, L2HGDH | |
| Mouse | |
| Protein A | |
| RUO | |
| 100152180, 79944 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction