missing translation for 'onlineSavingsMsg'
Learn More

L2HGDH Mouse anti-Human, Porcine, Clone: 3D11C2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66269-1-IG-20UL

 View more versions of this product

Product Code. 16807995

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes L-2-hydroxyglutarate dehydrogenase, a FAD-dependent enzyme that oxidizes L-2-hydroxyglutarate to alpha-ketoglutarate in a variety of mammalian tissues. Mutations in this gene cause L-2-hydroxyglutaric aciduria, a rare autosomal recessive neurometabolic disorder resulting in moderate to severe mental retardation.
Specifications

Specifications

L2HGDH
Monoclonal
1.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H9P8
L2HGDH
L2HGDH Fusion Protein Ag8382
20 μL
Primary
Human, Pig
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
3D11C2
Unconjugated
L2HGDH
C14orf160, DURANIN, L2HGDH
Mouse
Protein A
RUO
100152180, 79944
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.