missing translation for 'onlineSavingsMsg'
Learn More

L1TD1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18433435 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433435 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433435 Supplier Novus Biologicals Supplier No. NBP281960MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

L1TD1 Polyclonal antibody specifically detects L1TD1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen L1TD1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ECAT11, ES cell-associated protein 11, FLJ10884, LINE-1 type transposase domain containing 1, LINE-1 type transposase domain-containing protein 1, MGC133253, RP5-1155K23.3
Host Species Rabbit
Immunogen L1TD1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human L1TD1. The immunogen is located within amino acids 690 - 740 of L1TD1. Amino Acid Squence: IPEKESYENRAEDIIKE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54596
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.