missing translation for 'onlineSavingsMsg'
Learn More

L1CAM Antibody (S04-8I3), Novus Biologicals™

Rabbit Monoclonal Antibody

£328.00 - £501.00

Specifications

Antigen L1CAM
Clone S04-8I3
Concentration 0.3 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18603024
View Documents
Novus Biologicals
NBP3-20043-100ul
100 μg
£501.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18663735
View Documents
Novus Biologicals
NBP3-20043-50ul
50 μg
£328.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Western Blot
Specifications

Specifications

L1CAM
0.3 mg/mL
Monoclonal
Purified
RUO
Human
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
Recombinant protein of human L1CAM (Uniprot # P32004)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
S04-8I3
Western Blot
Unconjugated
Rabbit
Cellular Markers, Glia Markers, Neuroscience, Phospho Specific
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
3897
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.