missing translation for 'onlineSavingsMsg'
Learn More

L1CAM Antibody (CL12984), Novus Biologicals™

Mouse Monoclonal Antibody

£338.00 - £482.00

Specifications

Antigen L1CAM
Clone CL12984
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18658294
View Documents
Novus Biologicals
NBP3-18568-100ul
100 μg
£482.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18691315
View Documents
Novus Biologicals
NBP3-18568-25ul
25 μg
£338.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

L1CAM
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Monoclonal
Purified
RUO
Human
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
This antibody was developed against a sythetic peptide for P32004. Actual immunogen sequence is proprietary.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
CL12984
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cellular Markers, Glia Markers, Neuroscience, Phospho Specific
PBS, pH 7.2, 40% glycerol
3897
IgG2a
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.