missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1CAM Antibody (2702C), PE/Atto594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB7773PEATT594
This item is not returnable.
View return policy
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Flow Cytometry
Specifications
| L1CAM | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2702C | |
| Flow Cytometry | |
| antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| Mouse myeloma cell line NS0 derived Human L1CAM, (Ile20Glu1120) and (Arg864Glu1120), Accession # CAA42508 | |
| 0.1 mL | |
| Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
| 3897 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction