missing translation for 'onlineSavingsMsg'
Learn More

L1CAM Antibody (014), HRP, Novus Biologicals™

Product Code. 18392240 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392240 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392240 Supplier Novus Biologicals Supplier No. NBP289294H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

L1CAM Monoclonal specifically detects L1CAM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L1CAM
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Monoclonal
Clone 14
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Detection
Formulation PBS
Gene Alias antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human L1CAM (NP_000416.1; Met1-Glu1120).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Glia Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3897
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.