missing translation for 'onlineSavingsMsg'
Learn More

L189, MedChemExpress

Product Code. 16472737
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472737 50 mg 50mg
16436658 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472737 Supplier Medchem Express Supplier No. HY15588

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L189 is a novel human DNA ligase inhibitor, inhibits hLigI/III/IV with IC50 of 5/9/5 (mu)M.

L189 is a novel human DNA ligase inhibitor, inhibits hLigI/III/IV with IC50 of 5/9/5 (mu)M.

Specifications

Chemical Name or Material L189
CAS 64232-83-3
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C11H10N4OS
Quantity 50 mg
Solubility Information DMSO : ≥ 33 mg/mL (133.99 mM)
SMILES OC1=NC(S)=NC(N)=C1/N=C/C2=CC=CC=C2
Molecular Weight (g/mol) 246.29
Formula Weight 246.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.