Learn More
Tocris Bioscience™ L189
Product Code. p-5545854
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 18014222
Brand: Tocris Bioscience™ 3561/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
DNA ligase I, III and IV inhibitor
L189 is a DNA ligase I, III and IV inhibitor (IC50 values are 5, 9 and 5 μM respectively) that blocks DNA binding (Ki = 5 μM for DNA ligase I). Preferentially inhibits step 2 of the ligation reaction, and inhibits base excision repair (BER) and non-homologous end joining (NHEJ). Specifically sensitizes cancer cells to DNA damage and increases the cytotoxicity of DNA-damaging agents.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Product Type L189
Molecular Weight (g/mol) 246.29
Inhibitors L189