Learn More
Tocris Bioscience™ L189
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
L189 is a DNA ligase I, III and IV inhibitor (IC50 values are 5, 9 and 5 μM respectively) that blocks DNA binding (Ki = 5 μM for DNA ligase I). Preferentially inhibits step 2 of the ligation reaction, and inhibits base excision repair (BER) and non-homologous end joining (NHEJ). Specifically sensitizes cancer cells to DNA damage and increases the cytotoxicity of DNA-damaging agents.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | L189 |
| Molecular Weight (g/mol) | 246.29 |
| Inhibitors | L189 |